Skip to Content
Merck
All Photos(1)

Key Documents

803561

Sigma-Aldrich

BMOE (bis-maleimidoethane)

Synonym(s):

1,1′-ethane-1,2-diylbis(1H-pyrrole-2,5-dione), N,N′-Ethylenebismaleimide, BMOE, Ethylenebismaleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O4
CAS Number:
Molecular Weight:
220.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder

mol wt

220.18

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

DMSO or DMF: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(C=CC1=O)N1CCN2C(C=CC2=O)=O

InChI

1S/C10H8N2O4/c13-7-1-2-8(14)11(7)5-6-12-9(15)3-4-10(12)16/h1-4H,5-6H2

InChI key

PUKLCKVOVCZYKF-UHFFFAOYSA-N

General description

BMOE, BMB and BMH are homobifunctional, maleimide crosslinkers for conjugation between sulfhydryl groups (-SH). Such bismaleimide crosslinkers are commonly used to explore and characterize protein structure (i.e., oligomerization) or protein interactions. Because BMOE, BMB and BMH have the same reactivity but differ in length, the relative success of these three reagents in forming crosslinks between sites in a protein oligomer or interaction can assist in determining intra- and intermolecular distances.

Features and Benefits

  • Reactive groups: maleimide (both ends)
  • Reactive towards: sulfhydryl groups
  • Short (8.0Å), sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 7-atom spacer arm
  • Shortest bismaleimide crosslinker available for close proximity crosslinking
  • Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
  • Noncleavable

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M A Stalteri et al.
Bioconjugate chemistry, 6(2), 179-186 (1995-03-01)
Cross-linked F(ab')2 fragments derived from PR1A3, a murine monoclonal antibody used in radioimmunoscintigraphy of colorectal tumors, were produced using the bifunctional reagent bismaleimidohexane (BMH) as follows: Digestion of PR1A3 with pepsin gave F(ab')2 fragments which were purified by ion-exchange chromatography.
Z Xie et al.
The Journal of biological chemistry, 275(34), 25959-25964 (2000-06-15)
The Vibrio parahaemolyticus sodium/glucose transporter (vSGLT) is a bacterial member of the SGLT gene family. Wild-type and mutant vSGLT proteins were expressed in Escherichia coli, and transport activity was measured in intact cells and plasma membrane vesicles. Two cysteine-less vSGLT
Yong-Kook Kwon et al.
The Journal of cell biology, 163(2), 375-384 (2003-10-22)
A thiol-reactive membrane-associated protein (TRAP) binds covalently to the cytoplasmic domain of the human insulin receptor (IR) beta-subunit when cells are treated with the homobifunctional cross-linker reagent 1,6-bismaleimidohexane. Here, TRAP was found to be phospholipase C gamma1 (PLCgamma1) by mass
Kyoko Takebe et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(14), 2037-2047 (2003-11-05)
Epimorphin is a mesenchymal morphogen that has been shown to mediate epithelial-mesenchymal signaling interactions in various organs. We now show that epimorphin functions in hair follicle morphogenesis; using a novel ex vivo organ culture assay, we define a mechanism for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service