Skip to Content
Merck
All Photos(2)

Key Documents

802956

Sigma-Aldrich

2-Ethynylpyrimidine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€144.00

€144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€144.00

About This Item

Empirical Formula (Hill Notation):
C6H4N2
CAS Number:
Molecular Weight:
104.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€144.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

96.5 °C

SMILES string

C#CC1=NC=CC=N1

InChI

1S/C6H4N2/c1-2-6-7-4-3-5-8-6/h1,3-5H

InChI key

BMNUHRHTDNKJII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service