Skip to Content
Merck
All Photos(1)

Key Documents

802107

Sigma-Aldrich

Poly(N-isopropyl acrylamide), amine terminated

end functionalized by RAFT, PDI ≤1.3, Mn 5,000

Synonym(s):

NIPAM polymer, PNIPAM-NH2, functionalized polyNIPAM, functionalized polyacrylamide, polyacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(C6H11NO)n(CH2)NH2
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

Mn 5,000

PDI

≤1.3

Looking for similar products? Visit Product Comparison Guide

Application

Novel polymer for development of thermosensitive coated micro/nano materials including thermoresponsive polymeric drug delivery systems. The amine functinal group can be used to attach a variety of biomolecules to the polymer chain.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents: WO98/01478, WO99/311444.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reversibly thermo-responsive alkyl-terminated poly(N-isopropylacrylamide) core-shell micellar structures.
Chung JE, et al.
Colloids and Surfaces. B, Biointerfaces, 9(1-2), 37-48 (1997)
Hua Wei et al.
Biomaterials, 28(1), 99-107 (2006-09-09)
A four-arm star block copolymer, comprised of a hydrophobic PMMA arm and an average of three hydrophilic poly(N-isopropylacrylamide) (PNIPAAm) arms were designed and synthesized from the molecular level. The amphiphilic star block copolymer is capable of self-assembling into micelles in

Articles

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service