Skip to Content
Merck
All Photos(1)

Key Documents

799548

Sigma-Aldrich

L-Lactic acid-3-13C

greener alternative

99 atom % 13C, 98% (CP), ≥99% (Chiral Purity)

Synonym(s):

2-Hydroxypropanoic acid-3-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€3,460.00

€3,460.00


Estimated to ship on24 May 2025


Request more information

Select a Size

Change View
1 G
€3,460.00

About This Item

Linear Formula:
13CH3CH(OH)CO2H
CAS Number:
Molecular Weight:
91.07
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€3,460.00


Estimated to ship on24 May 2025


Request more information

isotopic purity

99 atom % 13C

Assay

≥99% (Chiral Purity)
98% (CP)

form

solid

greener alternative product score

old score: 45
new score: 14
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

bio NMR: suitable
mass spectrometry (MS): suitable

greener alternative category

mass shift

M+1

shipped in

dry ice

storage temp.

−20°C

SMILES string

[13CH3][C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1/i1+1

InChI key

JVTAAEKCZFNVCJ-IJGDANSWSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses”, “Design for Energy Efficiency” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service