Skip to Content
Merck
All Photos(1)

Key Documents

799505

Sigma-Aldrich

Ethane diamine cycloimine organic cage

Synonym(s):

CC1, Cage 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H48N12
CAS Number:
Molecular Weight:
792.98
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

>98%

form

powder

storage temp.

2-8°C

SMILES string

C1(/C=N/CC/N=C/C2=CC(/C=N/CC/N=C/3)=CC(/C=N/CC/N=C/4)=C2)=CC(/C=N/CC/N=C/C5=CC4=CC6=C5)=CC(/C=N/CC/N=C/C7=CC3=CC(/C=N/CC/N=C/6)=C7)=C1

InChI

1S/C48H48N12/c1-2-50-26-38-15-41-18-42(16-38)30-54-8-10-58-34-46-20-44-22-48(24-46)36-60-12-11-59-35-47-21-43(19-45(23-47)33-57-9-7-53-29-41)31-55-5-3-51-27-39-13-37(25-49-1)14-40(17-39)28-52-4-6-56-32-44/h13-36H,1-12H2/b49-25+,50-26+,51-27+,52-28+,53-29+,54-30+,55-31+,56-32+,57-33+,58-34+,59-35+,60-36+

InChI key

GUTZSUDNSFKJNV-WMKISWKQSA-N

Application

Cage-MOFs are metal organic frameworks (MOFs) having porous organic ligands. The CC1 cage finds applications as organic linker for ‘Cage-MOFs′ synthesis.[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shashikala I Swamy et al.
Journal of the American Chemical Society, 132(37), 12773-12775 (2010-08-28)
We report here the synthesis of a metal-organic framework comprising an organic cage linker with covalently prefabricated, intrinsic porosity. The network can be compared to a porous rock salt structure where the pores are partially filled by charge-balancing cations.
Tomokazu Tozawa et al.
Nature materials, 8(12), 973-978 (2009-10-27)
Porous materials are important in a wide range of applications including molecular separations and catalysis. We demonstrate that covalently bonded organic cages can assemble into crystalline microporous materials. The porosity is prefabricated and intrinsic to the molecular cage structure, as
Jamie L Culshaw et al.
Journal of the American Chemical Society, 135(27), 10007-10010 (2013-06-22)
A simple, one-step amidation reaction is used to produce a range of 12-arm organic building blocks for supramolecular chemistry via the derivatization of porous imine cages. As an example, microporous dendrimers are prepared.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service