Skip to Content
Merck
All Photos(1)

Key Documents

798215

Sigma-Aldrich

L-Fmoc-4-fluorophenylalanine

Synonym(s):

Fmoc-Phe(4-F)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-4-fluoro-L-phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€67.30

€67.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€67.30

About This Item

Empirical Formula (Hill Notation):
C24H20FNO4
CAS Number:
Molecular Weight:
405.42
Beilstein:
8287377
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€67.30


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(F)cc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H20FNO4/c25-16-11-9-15(10-12-16)13-22(23(27)28)26-24(29)30-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,26,29)(H,27,28)/t22-/m0/s1

InChI key

IXUMACXMEZBPJG-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service