Skip to Content
Merck
All Photos(1)

Key Documents

797529

Sigma-Aldrich

tert-Butyl 3-hydroxy-1-oxa-6-azaspiro[3.3]heptane-6-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

liquid

functional group

ether
hydroxyl

storage temp.

2-8°C

SMILES string

OC1C2(CN(C(OC(C)(C)C)=O)C2)OC1

InChI

1S/C10H17NO4/c1-9(2,3)15-8(13)11-5-10(6-11)7(12)4-14-10/h7,12H,4-6H2,1-3H3

InChI key

BFXOBAHLWARETK-UHFFFAOYSA-N

Application

Spirocyclic building block is part of a series of advanced angular[3.3]heptanes developed by Carreira and coworkers

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service