Skip to Content
Merck
All Photos(1)

Documents

797340

Sigma-Aldrich

1,2-Bis[4-(azidomethyl)phenyl]-1,2-diphenylethene

Synonym(s):

TPE-MN3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22N6
CAS Number:
Molecular Weight:
442.51
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

SMILES string

[N-]=[N+]=NCC(C=C1)=CC=C1/C(C2=CC=CC=C2)=C(C3=CC=CC=C3)/C4=CC=C(CN=[N+]=[N-])C=C4

InChI

1S/C28H22N6/c29-33-31-19-21-11-15-25(16-12-21)27(23-7-3-1-4-8-23)28(24-9-5-2-6-10-24)26-17-13-22(14-18-26)20-32-34-30/h1-18H,19-20H2/b28-27+

InChI key

IXQXFSMBYFNNPN-BYYHNAKLSA-N

Application

TPE-MN3 is an aggregation-induced emission (AIE) dye for "Click" reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-Free Click Polymerization: Synthesis and Photonic Properties of Poly(aroyltriazole)s
Qin et al.
Advances in Functional Materials, 19, 1891?1900-1891?1900 (2009)

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service