Skip to Content
Merck
All Photos(1)

Key Documents

796859

Sigma-Aldrich

2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane

95%

Synonym(s):

Cyclohexadimethylsiloxane, Dodecamethylcyclohexasiloxane, Dodecamethylhexacyclosiloxane, Dow Corning 246

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€63.60

€63.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€63.60

About This Item

Empirical Formula (Hill Notation):
C12H36O6Si6
CAS Number:
Molecular Weight:
444.92
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€63.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

SMILES string

C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

InChI

1S/C12H36O6Si6/c1-19(2)13-20(3,4)15-22(7,8)17-24(11,12)18-23(9,10)16-21(5,6)14-19/h1-12H3

InChI key

IUMSDRXLFWAGNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane(Dodecamethylcyclohexasiloxane (D6)) is widely used in the chemical industry. It is mainly used as a raw material for silicone gel, rubber, and resin [1]. It undergoes ring opening polymerization to obtain polydodecamethylcyclohexasiloxane polymer.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exposure to dodecamethylcyclohexasiloxane (D6) affects the antioxidant response and gene expression of Procambarus clarkii
Hossain MM, et al.
Sustainability, 13(6), 3495-3495 (2021)
Maghnite-H+ as inorganic acidic catalyst in ring opening polymerization of dodecamethylcyclohexasiloxane
Kherroub DE, et al.
Silicon, 11(2), 1165-1173 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service