Skip to Content
Merck
All Photos(1)

Key Documents

79505

Sigma-Aldrich

Trimethyl phosphonoformate

≥97.0%

Synonym(s):

Methyl dimethoxyphosphinylformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)CO2CH3
CAS Number:
Molecular Weight:
168.09
Beilstein:
1770919
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0%

refractive index

n20/D 1.423 (lit.)
n20/D 1.423

bp

50-52 °C/0.02 mmHg

density

1.254 g/mL at 25 °C (lit.)

SMILES string

COC(=O)P(=O)(OC)OC

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service