Skip to Content
Merck
All Photos(3)

Key Documents

793507

Sigma-Aldrich

Methylammonium bromide

greener alternative

98%

Synonym(s):

Methanamine hydrobromide, Methylamine, hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€221.00

€221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€221.00

About This Item

Linear Formula:
CH3NH2 · HBr
CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

€221.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

255-260 °C

greener alternative category

SMILES string

CN.Br

InChI

1S/CH5N.BrH/c1-2;/h2H2,1H3;1H

InChI key

ISWNAMNOYHCTSB-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Methylamine hydobromide is an important precursor for the preparation of perovskite photoactive layers for solar energy conversion[1].
Methylammonium bromide (MABr) can be used as a precursor in the preparation of methylammonium lead bromide based perovskite material with good optical properties, which include green emission, and photoluminescence. This material can further be utilized in the fabrication of renewable energy devices such as light emitting diodes(LEDs), perovskite solar cells (PSCs), and photovoltaic cells.[2][3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced mobility CsPbI3 quantum dot arrays for record-efficiency, high-voltage photovoltaic cells
Sanehira EM, et al.
Science advances, 3(10), eaao4204-eaao4204 (2017)
Optical and electronic properties of mixed halide (X= I, Cl, Br) methylammonium lead perovskite solar cells
Tombe S, et al.
Journal of Material Chemistry C, 5(7), 1714-1723 (2017)
Michael M Lee et al.
Science (New York, N.Y.), 338(6107), 643-647 (2012-10-09)
The energy costs associated with separating tightly bound excitons (photoinduced electron-hole pairs) and extracting free charges from highly disordered low-mobility networks represent fundamental losses for many low-cost photovoltaic technologies. We report a low-cost, solution-processable solar cell, based on a highly

Articles

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service