Skip to Content
Merck

Skip To

791105

Zinc chloromethanesulfinate

95% (H-NMR)

Synonym(s):

Baran MCMS Reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 618,00

About This Item

Empirical Formula (Hill Notation):
C2H4Cl2O4S2Zn
CAS Number:
Molecular Weight:
292.48
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352101
MDL number:
Assay:
95% (H-NMR)
Form:
solid

€ 618,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

95% (H-NMR)

form

solid

reaction suitability

reaction type: C-C Bond Formation, reagent type: catalyst
reaction type: C-H Activation, reagent type: diversification reagent

mp

190-195 °C

functional group

chloro, sulfinic acid

storage temp.

2-8°C

SMILES string

ClCS(O[Zn]OS(CCl)=O)=O

InChI

1S/2CH3ClO2S.Zn/c2*2-1-5(3)4;/h2*1H2,(H,3,4);/q;;+2/p-2

InChI key

HEKATMPPXNXYJU-UHFFFAOYSA-L

Application

Zinc chloromethanesulfinate (MCMS) is part of a zinc sulfinate toolbox developed by Phil Baran for the simple and rapid diversification of heterocycles.1

Practical and Innate Carbon-Hydrogen Functionalization of Heterocycles

Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
745480771406746118
assay

95% (H-NMR)

assay

95%

assay

-

assay

95%

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

reaction suitability

reaction type: C-C Bond Formation, reagent type: diversification reagent, reagent type: catalyst
reaction type: C-H Activation

reaction suitability

reaction type: Allylation, reaction type: C-C Bond Formation, reagent type: catalyst
reaction type: C-H Activation, reagent type: diversification reagent

reaction suitability

reaction type: C-C Bond Formation, reagent type: catalyst
reaction type: C-H Activation

reaction suitability

reaction type: C-C Bond Formation, reagent type: catalyst
reaction type: C-H Activation, reagent type: linker

form

solid

form

solid

form

solid

form

solid

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

mp

190-195 °C

mp

161-166 °C

mp

151-157 °C

mp

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters