Skip to Content
Merck
All Photos(1)

Key Documents

790753

Sigma-Aldrich

(R)-3,3′-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1′-binaphthyl-2,2′-disulfonimide

95%

Synonym(s):

(11bR)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]dinaphtho[2,1-d:1′,2′-f][1,3,2]dithiazepine 3,3,5,5-tetraoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H17F12NO4S2
CAS Number:
Molecular Weight:
819.64
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]22/D -42.0°, c = 0.5% in chloroform

reaction suitability

reaction type: Aldol Reaction
reaction type: Allylation
reaction type: Carbonyl/Imine Addition
reaction type: Friedel-Crafts Alkylation

mp

325-330 °C

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)-c2cc3ccccc3c4-c5c(c(cc6ccccc56)-c7cc(cc(c7)C(F)(F)F)C(F)(F)F)S(=O)(=O)NS(=O)(=O)c24

InChI

1S/C36H17F12NO4S2/c37-33(38,39)21-9-19(10-22(15-21)34(40,41)42)27-13-17-5-1-3-7-25(17)29-30-26-8-4-2-6-18(26)14-28(32(30)55(52,53)49-54(50,51)31(27)29)20-11-23(35(43,44)45)16-24(12-20)36(46,47)48/h1-16,49H

InChI key

DFXCMPXSCANWFA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Asymmetric counteranion-directed catalysis (ACDC) has a number of valuable applications in enantioselective synthesis.

Related Content

The List group focuses on the development of new concepts in catalysis. Since 1999, this research group has pioneered the development of organocatalysis as the third pillar of stereoselective catalysis, along side biocatalysis and transition metal catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service