Skip to Content
Merck
All Photos(1)

Documents

790567

Sigma-Aldrich

2-Tetrahydropyranyl acrylate

contains <500 ppm inhibitor, ≥97%

Synonym(s):

2-Propenoic acid tetrahydro-2H-pyran-2-yl ester, THPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O3
CAS Number:
Molecular Weight:
156.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

form

liquid

contains

<500 ppm inhibitor

refractive index

n20/D 1.457

density

1.05 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC(OC1OCCCC1)=O

InChI

1S/C8H12O3/c1-2-7(9)11-8-5-3-4-6-10-8/h2,8H,1,3-6H2

InChI key

FGWRVVZMNXRWDQ-UHFFFAOYSA-N

Application

This acrylate is commonly used in the formation of photoresists, but can also be used as a protected acrylic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.0 °F

Flash Point(C)

80.55 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mattia Sanna et al.
Tobacco control, 29(1), 36-42 (2018-11-07)
Adult smoking prevalence in Taiwan rapidly declined from 26.5% in 2005 to 20.0% in 2015. Nevertheless, future projections on smoking-attributable deaths and current per capita consumption do not paint an equally bright picture. We used SimSmoke, a tobacco control simulation
Ignacio Lacalle et al.
Sensors (Basel, Switzerland), 20(15) (2020-07-30)
During the past few decades, the combination of flourishing maritime commerce and urban population increases has made port-cities face several challenges. Smart Port-Cities of the future will take advantage of the newest IoT technologies to tackle those challenges in a
Jiaxin Liu et al.
Cancer cell international, 20, 342-342 (2020-08-04)
Aberrant methylation and miRNA-target-gene regulation function as important mechanisms for gene inactivation in colon carcinogenesis. Although a serious of molecular events (such as aberrant alterations of genomics and epigenetics) have been identified to be related to prognostic in colon cancer
Petzetakis; N.;
PMSE Preprints (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service