Skip to Content
Merck
All Photos(1)

Key Documents

78727

Sigma-Aldrich

2-Phenyl-2-imidazoline

≥96% (HPLC)

Synonym(s):

4,5-Dihydro-2-phenyl-1H-imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€57.40

€57.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€57.40

About This Item

Empirical Formula (Hill Notation):
C9H10N2
CAS Number:
Molecular Weight:
146.19
Beilstein:
119250
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥96% (HPLC)

form

flakes

SMILES string

C1CN=C(N1)c2ccccc2

InChI

1S/C9H10N2/c1-2-4-8(5-3-1)9-10-6-7-11-9/h1-5H,6-7H2,(H,10,11)

InChI key

BKCCAYLNRIRKDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • N-arylation of indoles
  • Epoxy-carbonyl reactions

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

393.8 °F - closed cup

Flash Point(C)

201 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 2-phenyl-2-imidazoline in air by high-performance liquid chromatography with ultraviolet detection.
J Pukkila et al.
Journal of chromatography, 438(2), 443-446 (1988-04-22)
Komal Kumar et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-16)
Levulinic acid (LA) is an industrially important product that can be catalytically valorized into important value-added chemicals. In this study, hydrothermal conversion of glucose into levulinic acid was attempted using Brønsted acidic ionic liquid catalyst synthesized using 2-phenyl-2-imidazoline, and 2-phenyl-2-imidazoline-based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service