Skip to Content
Merck
All Photos(1)

Key Documents

778575

Sigma-Aldrich

(S)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthalene

Synonym(s):

(S)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16Br2O2
CAS Number:
Molecular Weight:
472.17
Beilstein:
4712473
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

SMILES string

COc1c(Br)cc2ccccc2c1-c3c(OC)c(Br)cc4ccccc34

InChI

1S/C22H16Br2O2/c1-25-21-17(23)11-13-7-3-5-9-15(13)19(21)20-16-10-6-4-8-14(16)12-18(24)22(20)26-2/h3-12H,1-2H3

InChI key

DFTUKDIMHCCQIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service