Skip to Content
Merck
All Photos(2)

Key Documents

773107

Sigma-Aldrich

Fmoc-N-Me-Lys(Boc)-OH

97% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€486.00
1 G
€1,320.00

€486.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€486.00
1 G
€1,320.00

About This Item

Empirical Formula (Hill Notation):
C27H34N2O6
CAS Number:
Molecular Weight:
482.57
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€486.00


Check Cart for Availability

Request a Bulk Order

Assay

97% (HPLC)

form

powder or crystals

optical activity

[α]22/D -14.0°, c = 1% in chloroform

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

189-194 °C (decomposition)

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

N([C@@H](CCCCNC(=O)OC(C)(C)C)C(=O)O)(C)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C27H34N2O6/c1-27(2,3)35-25(32)28-16-10-9-15-23(24(30)31)29(4)26(33)34-17-22-20-13-7-5-11-18(20)19-12-6-8-14-21(19)22/h5-8,11-14,22-23H,9-10,15-17H2,1-4H3,(H,28,32)(H,30,31)/t23-/m0/s1

InChI key

JMBKBGOKNZZJQA-QHCPKHFHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service