Skip to Content
Merck
All Photos(1)

Documents

769800

Sigma-Aldrich

Resomer® RG 753 S, Poly(D,L-lactide-co-glycolide)

ester terminated, Lactide: Glycolide 75:25, lactide:glycolide 75:25

Synonym(s):

Poly(Glycolide Lactide)Copolymer, Poly(Lactide-Co-Glycolide), Resomer® RG 753 S, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[C2H4O2]n[C2H2O2]mCH3
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

amorphous

feed ratio

lactide:glycolide 75:25

viscosity

0.32-0.44 dL/g

transition temp

Tg 44-50 °C

storage temp.

2-8°C

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Resomer® RG 753 S, Poly(D,L-lactide-co-glycolide) is a biodegradable and biocompatible aliphatic polyester, which can be used in the development of nano and microparticles. It is a pharmaceutical polymer that can be potentially utilized in drug delivery and gene delivery. It forms a three-dimensional porous scaffold, which can be used in tissue engineering.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and evaluation of novel biodegradable microspheres based on poly (d, l-lactide-co-glycolide) and poly (varepsilon-caprolactone) for controlled delivery of doxycycline in the treatment of human periodontal pocket: in vitro and in vivo studies
Mundargi RC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 119(1), 59-68 (2007)
Nanoparticles made of fluorescence-labelled poly (L-lactide-co-glycolide): preparation, stability, and biocompatibility
Weiss B, et al.
Journal of Nanoscience and Nanotechnology, 6(9-10), 3048-3056 (2006)
Nano/micro technologies for delivering macromolecular therapeutics using poly (D, L-lactide-co-glycolide) and its derivatives
Mundargi RC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 125(3), 193-209 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service