Skip to Content
Merck
All Photos(1)

Key Documents

768944

Sigma-Aldrich

4-Pentyn-1-amine hydrochloride

≥97.0% (GC)

Synonym(s):

1-Amino-4-pentyne hydrochloride, 4-Pentynylamine hydrochloride, 5-Amino-1-pentyne hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€248.00

€248.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€248.00

About This Item

Empirical Formula (Hill Notation):
C5H9N· HCl
CAS Number:
Molecular Weight:
119.59
Beilstein:
7376466
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€248.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)
97.0-103.0%

form

powder

functional group

amine

SMILES string

C#CCCCN.Cl

InChI

1S/C5H9N.ClH/c1-2-3-4-5-6;/h1H,3-6H2;1H

InChI key

AJKFBGIRKUYQBB-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service