Skip to Content
Merck
All Photos(1)

Key Documents

768936

Sigma-Aldrich

(S)-1,1-Dimethoxy-2-propanol

≥99.0% (sum of enantiomers, GC)

Synonym(s):

(2S)-2-Hydroxypropionaldehyde dimethyl acetal, (S)-1,1-Dimethoxy-2-hydroxypropane, (S)-Lactaldehyde dimethyl acetal

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€426.00
5 G
€1,800.00

€426.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€426.00
5 G
€1,800.00

About This Item

Empirical Formula (Hill Notation):
C5H12O3
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

€426.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

form

liquid

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

functional group

acetal
ether
hydroxyl

storage temp.

2-8°C

SMILES string

C[C@H](O)C(OC)OC

InChI

1S/C5H12O3/c1-4(6)5(7-2)8-3/h4-6H,1-3H3/t4-/m0/s1

InChI key

PRAYXKGWSGUXQK-BYPYZUCNSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.3 °F

Flash Point(C)

50.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service