Skip to Content
Merck
All Photos(1)

Key Documents

768766

Sigma-Aldrich

9,9′-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis-9H-carbazole

98%

Synonym(s):

DMFL-CBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H28N2
CAS Number:
Molecular Weight:
524.65
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

280-286 °C

λmax

293, 341 nm in THF

fluorescence

λem 363 nm in THF

SMILES string

CC1(C)c2ccc(cc2-c3cc(ccc13)-n4c5ccccc5c6ccccc46)-n7c8ccccc8c9ccccc79

InChI

1S/C39H28N2/c1-39(2)33-21-19-25(40-35-15-7-3-11-27(35)28-12-4-8-16-36(28)40)23-31(33)32-24-26(20-22-34(32)39)41-37-17-9-5-13-29(37)30-14-6-10-18-38(30)41/h3-24H,1-2H3

InChI key

XWUMQFHANBCENC-UHFFFAOYSA-N

Application

This material is a popular phopshorescent host material for OLED devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service