Skip to Content
Merck
All Photos(1)

Key Documents

767530

Sigma-Aldrich

Poly(ethylene glycol) bisazide

average Mn 10,000, azide

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002

Product Name

Poly(ethylene glycol) bisazide, average Mn 10,000

form

solid

mol wt

PEG average Mn 10,000 (n~230)
average Mn 10,000

transition temp

Tm 59-64 °C

Ω-end

azide

α-end

azide

storage temp.

2-8°C

Application

PEG functionalized with azide for ligation and coupling reactions. This can also be crosslinked into hydrogels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service