Skip to Content
Merck
All Photos(1)

Key Documents

765414

Sigma-Aldrich

Gold nanoparticles

50 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O

Synonym(s):

Gold nanoparticles NH2 functionalized, Au NP NH2, Gold Colloid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

material

PEG 5000

Quality Level

form

dispersion in H2O
nanoparticles

packaging

poly bottle of 1 mL

OD

50

diameter

50 nm

pH

6.0-8.0 (25 °C)

solubility

water: miscible

density

1.00 g/cm3

λmax

535 nm

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Amine functionalized GNPs which has Poly Ethylene Glycol (PEG) as the ligand or carrier molecule forms a conjugate with siRNA and is extremely useful in RNAi technology.

Application

Functionalized particles have been extensively used to study cellular uptake of nanoparticles and targeted drug delivery.
Gold nanoparticles 50 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O can be used in the activation of silica nanosphere for the decomposition of formic acid. It may be used in the radiosensitization and imaging of cancer cells.
AuNPs can be potentially immobilized on antibodies to improve the sensitivity of optical fiber sensors for the screening of COVID-19. It can also be deposited on the titanium electrode for the formation of a miniaturized device for the electrochemical detection of DNA hybridization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical Fiber Sensors for Rapid Screening of COVID-19
Nag P, et al.
Transactions of the Indian National Academy of Engineering, 152(25), 1-4 (2020)
Strong metal-molecular support interaction (SMMSI): amine-functionalized gold nanoparticles encapsulated in silica nanospheres highly active for catalytic decomposition of formic acid
Yadav M, et al.
Journal of Materials Chemistry, 22(25), 12582-12586 (2012)
Gold nanoparticles for radiosensitizing and imaging of cancer cells
Borran AA, et al.
Radiation Physics and Chemistry, 152(25), 137-144 (2018)
Scott H Brewer et al.
Langmuir : the ACS journal of surfaces and colloids, 21(20), 9303-9307 (2005-09-21)
The interaction of bovine serum albumin (BSA) with gold colloids and surfaces was studied using zeta-potential and quartz crystal microbalance (QCM) measurements, respectively, to determine the surface charge and coverage. The combination of these two measurements suggests that BSA binding
Functionalized gold nanoparticles and their biomedical applications
Tiwari, P. M., Vig, K., Dennis, V. A., & Singh, S. R.
Nanomaterials, 1(1), 31-63 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service