Skip to Content
Merck
All Photos(3)

Key Documents

764531

Sigma-Aldrich

Cerium(IV) oxide-yttria doped

greener alternative

nanopowder, contains 10 mol % yttria, <50 nm particle size, 99.9% trace metals basis

Synonym(s):

10YDC, YDC-10, YDC10, Cerium yttrium oxide, YDC, Yttria doped ceria, Yttrium doped ceria

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2/Y2O3
CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

99.9% trace metals basis

form

nanopowder

contains

10 mol % yttria

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<50 nm

mp

>400 °C

greener alternative category

SMILES string

O=[Ce]=O.O=[Y]O[Y]=O

InChI

1S/Ce.5O.2Y

InChI key

XVLUKJGVKWVXMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

8mol% Yttria doped ceria may be prepared by carbonate coprecipitation technique. Yttria doped ceria exihibits high ionic conductivity, low activation energy and small grain boundary resistance. The effect of microstructure on the electrolytic properties was investigated in detail.
Cerium(IV) oxide-yttria doped (YDC-10) is an electrolytic material that is used as an alternative to zirconia-based electrolytes. Its properties include high ionic conductivity (0.01 S cm−1), electronic conductivity (3 S cm−1), enhanced electrode reactions, and high oxygen surface exchange coefficient.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

They are promising ceramic electrolytes for solid oxide fuel cells operating at low temperatures of 500-700°C (IT-SOFC). Doped ceria interlayers in between bulk yttria-stabilized zirconia electrolyte further enhance the performance of SOFCs.
YDC-10 can be used as an electrolyte in the fabrication of solid oxide fuel cells (SOFCs). A thin film of YDC-10 (~17.5 nm) effectively reduces the cathode/interfacial energy by 0.1 eV and increases the exchange current density by a factor of 4. YDC-10 can be operated with a temperature range of 300° - 500°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cales, B.; et al.
Journal of Physics and Chemistry of Solids, 45, 929-929 (1984)
Wang, Y; et al.
Solid State Ionics, 177, 1681-1681 (2006)
A Yitbarek et al.
Poultry science, 94(8), 1802-1811 (2015-06-08)
Prebiotics and synbiotics are considered to be among the most promising replacements for in-feed antibiotic growth promoters (AGP) in poultry feed. The current study was designed to study the effect of Bacitracin methylene disalicylate (BMD) (Control), yeast-derived carbohydrates (YDC), and
Fabrication and sintering of fine yttria-doped ceria powder
Van HJ, et al.
Journal of the American Ceramic Society. American Ceramic Society, 80(4), 933-940 (1997)
Synthesis, sintering and impedance spectroscopy of 8mol% yttria-doped ceria solid electrolyte.
Tadokoro SK, et al.
Journal of Power Sources, 130(1), 15-21 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service