Skip to Content
Merck
All Photos(2)

Key Documents

764256

Sigma-Aldrich

4-Iodo-2-methoxyphenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€89.40

€89.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€89.40

About This Item

Empirical Formula (Hill Notation):
C7H7IO2
CAS Number:
Molecular Weight:
250.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€89.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

37-40 °C

functional group

iodo

storage temp.

2-8°C

SMILES string

COc1cc(I)ccc1O

InChI

1S/C7H7IO2/c1-10-7-4-5(8)2-3-6(7)9/h2-4,9H,1H3

InChI key

KJXYWMSTOWBZHC-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Yeon Choi et al.
Neuromolecular medicine, 19(4), 555-570 (2017-10-21)
Alzheimer's disease (AD) is pathologically characterized by an excessive accumulation of amyloid-beta (Aβ) fibrils within the brain. We tested the anti-inflammatory and anti-amyloidogenic effects of (E)-2-methoxy-4-(3-(4-methoxyphenyl) prop-1-en-1-yl) phenol (MMPP), a selective signal transducer and activator of transcription 3 (STAT3) inhibitor.
Ji Yeon Choi et al.
International journal of molecular sciences, 20(11) (2019-05-31)
Neuroinflammation is implicated in dopaminergic neurodegeneration. We have previously demonstrated that (E)-2-methoxy-4-(3-(4-methoxyphenyl) prop-1-en-1-yl) phenol (MMPP), a selective signal transducer and activator of transcription 3 (STAT3) inhibitor, has anti-inflammatory properties in several inflammatory disease models. We investigated whether MMPP could protect
Florian Juhlke et al.
Frontiers in chemistry, 5, 120-120 (2018-01-13)
Chlorinated guaiacol derivatives are found in waste water of pulp mills using chlorine in the bleaching process of wood pulp. They can also be detected in fish tissue, possibly causing off-odors. To date, there is no systematic investigation on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service