Skip to Content
Merck
All Photos(1)

Key Documents

763233

Sigma-Aldrich

[2-(Dicyclohexylphosphino)-3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl]gold(I) bis(trifluoromethanesulfonyl)imide

Synonym(s):

BrettPhos AuNTf2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€181.00
1 G
€523.00

€181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€181.00
1 G
€523.00

About This Item

Empirical Formula (Hill Notation):
C37H53AuF6NO6PS2
CAS Number:
Molecular Weight:
1013.88
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€181.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: gold
reaction type: Cross Couplings
reagent type: catalyst

mp

192-209 °C (decomposition)

SMILES string

FC(F)(F)S(=O)(=O)N([Au])S(=O)(=O)C(F)(F)F.COc1ccc(OC)c(c1P(C2CCCCC2)C3CCCCC3)-c4c(cc(cc4C(C)C)C(C)C)C(C)C

InChI

1S/C35H53O2P.C2F6NO4S2.Au/c1-23(2)26-21-29(24(3)4)33(30(22-26)25(5)6)34-31(36-7)19-20-32(37-8)35(34)38(27-15-11-9-12-16-27)28-17-13-10-14-18-28;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/h19-25,27-28H,9-18H2,1-8H3;;/q;-1;+1

InChI key

SMTGSBLIUQSWAP-UHFFFAOYSA-N

Application

Bulky gold pi-acid catalyst used in oxidative cyclization reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youliang Wang et al.
Angewandte Chemie (International ed. in English), 52(30), 7795-7799 (2013-06-22)
Golden aryne? Gold aryne complexes are inferred as transition states in dual gold-catalyzed cyclizations of cis-enediynes (see scheme; DCE = 1,2-dichloroethane). They are better described as ortho-aurophenyl cations, which react with weak nucleophiles and undergo facile intramolecular insertions into C(sp(3))-H

Related Content

The major research thrust in Professor Liming Zhang’s group is homogeneous gold catalysis ranging from discovery of novel reactivities, design of new ligands, development of versatile synthetic methods and application in natural product synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service