Skip to Content
Merck
All Photos(1)

Key Documents

762407

Sigma-Aldrich

N-Boc-4-(2,2-difluorocyclopropyl)-L-proline

97%

Synonym(s):

6(S)-5-Boc-1,1-difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€292.00

€292.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€292.00

About This Item

Empirical Formula (Hill Notation):
C12H17F2NO4
Molecular Weight:
277.26
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€292.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

87-97 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CC2(C[C@H]1C(O)=O)CC2(F)F

InChI

1S/C12H17F2NO4/c1-10(2,3)19-9(18)15-6-11(5-12(11,13)14)4-7(15)8(16)17/h7H,4-6H2,1-3H3,(H,16,17)/t7-,11?/m0/s1

InChI key

OXXSWURHJYLTIL-RGENBBCFSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service