Skip to Content
Merck
All Photos(1)

Key Documents

760129

Sigma-Aldrich

2,5-Dibromo-3,4-dinitrothiophene

99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4Br2N2O4S
CAS Number:
Molecular Weight:
331.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
form:
solid
Assay:
99% (GC)

Assay

99% (GC)

form

solid

mp

135-140 °C

SMILES string

[O-][N+](=O)c1c(Br)sc(Br)c1[N+]([O-])=O

InChI

1S/C4Br2N2O4S/c5-3-1(7(9)10)2(8(11)12)4(6)13-3

InChI key

AHGHPBPARMANQD-UHFFFAOYSA-N

Application

This monomer is useful in the Pd catalyzed Stille coupling to make conjugated thiophene oligomers or co-oligomers. The corresponding amine(s) can be obtained by subsequent reduction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service