Skip to Content
Merck
All Photos(2)

Key Documents

757799

Sigma-Aldrich

Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether 2-(biotinylamino)ethane

average Mn 3,800, cross-linking reagent amine reactive, NHS ester, biotin

Synonym(s):

Polyethylene glycol, Biotin polyoxyethylene maleimide, Biotin-PEG, Biotin-PEG maleimide, Biotinylated PEG, NHS - protected acid PEG, Poly(ethylene glycol) (N-hydroxysuccinimide acetic acid) ether biotin, biotin carboxylic acid PEG, protected valeric acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€617.00

€617.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€617.00

About This Item

Linear Formula:
C10H16N3O2S(C2H4O)nC8H8NO5
UNSPSC Code:
12162002
NACRES:
NA.23

€617.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether 2-(biotinylamino)ethane, average Mn 3,800

form

solid

mol wt

PEG average Mn 3,400 (n ~ 77)
average Mn 3,800

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

mp

45-53 °C

Ω-end

NHS ester

α-end

biotin

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether 2-(biotinylamino)ethane is a heterobifunctional poly(ethylene glycol) (PEG) derivative with a biotin terminus. [1]

Application

bioconjugation, drug delivery, PEG hydrogel, crosslinker, surface functionalization

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basic studies on heterobifunctional biotin-PEG conjugates with a 3-(4-pyridyldithio) propionyl marker on the second terminus.
Kaiser K, et al.
Bioconjugate Chemistry, 8(4), 545-551 (1997)

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service