Skip to Content
Merck
All Photos(1)

Key Documents

75760

Sigma-Aldrich

Oxalyl chloride

dist., ≥96.0% (AT)

Synonym(s):

Ethanedioyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOCl
CAS Number:
Molecular Weight:
126.93
Beilstein:
1361988
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

4.4 (vs air)

vapor pressure

150 mmHg ( 20 °C)

Assay

≥96.0% (AT)

form

liquid

quality

dist.

reaction suitability

reagent type: oxidant

refractive index

n20/D 1.429 (lit.)

bp

62-65 °C (lit.)

mp

−10-−8 °C (lit.)

density

1.5 g/mL at 20 °C (lit.)

SMILES string

ClC(=O)C(Cl)=O

InChI

1S/C2Cl2O2/c3-1(5)2(4)6

InChI key

CTSLXHKWHWQRSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of N-heterocyclic ynones and ynediones, used to activate carboxylic acids
  • Chlorination and halogenation
  • Three-component [3+2] cycloadditions
  • Reactions with organostannanes
  • Synthesis of cyclopentenones
  • Carbonylations, used as a carbonyl synthon

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsutomu Kimura et al.
Chemical communications (Cambridge, England), (32)(32), 4077-4079 (2005-08-11)
Reaction of diastereomerically pure phosphinoselenoic acid salts with oxalyl chloride leads to enantiomerically pure P-chiral phosphinoselenoic chlorides with inversion of configuration at phosphorus; one of these chlorides is converted to a phosphinoselenothioic acid salt with a high degree of enantioselectivity.
Peter J Manley et al.
Organic letters, 4(18), 3127-3129 (2002-08-31)
[reaction: see text] A mild, practical, one-pot method for the generation of imidoyl chlorides and their subsequent in situ reaction with pyridine-1-oxides is described. The imidoyl chlorides were formed from the reaction of secondary amides with a stoichiometric amount of
Sham M Sondhi et al.
European journal of medicinal chemistry, 43(12), 2824-2830 (2007-11-21)
A series of substituted N-methylisonicotinamidine (2a-f), N-methylpyrazine-2-carboxamidine (2g-i) derivatives were synthesized by reaction of amidine derivatives (1a-i) with methyl iodide in presence of triethylamine. Five-membered condensed dihydroimidazolylbenzenesulfonamide derivatives (3a-i) were obtained by the reaction of amidine derivatives (1a-i) with acylating
Taisuke Itaya et al.
Chemical & pharmaceutical bulletin, 50(1), 83-86 (2002-02-05)
The relationship between the product patterns and the configurations of 1,2-cycloheptane- and 1,2-cyclooctanediols 9 in the cyclocondensations with oxalyl chloride in the presence of triethylamine at 0 degrees C has been shown analogous to that obtained for 1,2-disubstituted acyclic ethylene
Benoît Heurtaux et al.
The Journal of organic chemistry, 70(4), 1474-1477 (2005-02-12)
[reaction: see text] Several natural pulvinic acids were synthesized. Silyl ketene acetals derived from methyl arylacetates (4 equiv) reacted with oxalyl chloride at -78 degrees C, without the need of adding a catalyst. After treatment of the crude diketones with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service