Skip to Content
Merck
All Photos(1)

Key Documents

756911

Supelco

Triphenyl phosphate solution

NMR reference standard, 3 mM in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€74.10

€74.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
€74.10

About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

€74.10


Check Cart for Availability

Request a Bulk Order

grade

NMR reference standard
analytical standard

Assay

99% (CP)

concentration

3 mM in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

31P sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Cervera et al.
Analytical and bioanalytical chemistry, 406(27), 6843-6855 (2014-05-16)
An atmospheric pressure chemical ionization source has been used to enhance the potential of gas chromatography coupled with quadrupole time-of-flight (QTOF) mass spectrometry (MS) for screening and quantification purposes in pesticide residue analysis. A screening method developed in our laboratory
Katsumasa Abe et al.
Applied and environmental microbiology, 80(18), 5866-5873 (2014-07-20)
Phosphotriesterases catalyze the first step of organophosphorus triester degradation. The bacterial phosphotriesterases purified and characterized to date hydrolyze mainly aryl dialkyl phosphates, such as parathion, paraoxon, and chlorpyrifos. In this study, we purified and cloned two novel phosphotriesterases from Sphingomonas
Blagoj Mitrevski et al.
Journal of chromatography. A, 1362, 262-269 (2014-09-03)
Quadrupole time-of-flight mass spectrometry (Q-TOFMS) has been evaluated with respect to its applicability in comprehensive two-dimensional gas chromatography (GC×GC). At a maximum acquisition frequency, while approximately 50 full accurate mass spectra on disk were acquired per s (50Hz) in scan
Zulin Zhang et al.
Journal of separation science, 37(24), 3699-3705 (2014-10-02)
A simple and rapid method was developed for the simultaneous analysis of nine different pesticides in water samples by gas chromatography with mass spectrometry. A number of parameters that may affect the recovery of pesticides, such as the type of
Evgeni Eltzov et al.
Analytical chemistry, 87(7), 3655-3661 (2015-03-17)
Indoor air pollution became a recent concern found to be oftentimes worse than outdoor air quality. We developed a tool that is cheap and simple and enables continuous monitoring of air toxicity. It is a biosensor with both a nondisposable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service