Skip to Content
Merck
All Photos(2)

Key Documents

756377

Sigma-Aldrich

3-(2-Acetoxy-4,6-dimethylphenyl)-3-methylbutyric acid

97%

Synonym(s):

2-(Acetyloxy)-β,β,4,6-tetramethylbenzenepropanoic acid, 3-(2-Acetoxy-4,6-dimethylphenyl)-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€184.00
1 G
€872.00

€184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€184.00
1 G
€872.00

About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€184.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

113-118 °C

functional group

carboxylic acid
ester

storage temp.

2-8°C

SMILES string

CC(=O)Oc1cc(C)cc(C)c1C(C)(C)CC(O)=O

InChI

1S/C15H20O4/c1-9-6-10(2)14(12(7-9)19-11(3)16)15(4,5)8-13(17)18/h6-7H,8H2,1-5H3,(H,17,18)

InChI key

IRUYOPOSEOEWIN-UHFFFAOYSA-N

Application

Used as a trimethyl lock in stable chromogenic substrates for esterases[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael N Levine et al.
Molecules (Basel, Switzerland), 13(2), 204-211 (2008-02-29)
p-Nitrophenyl acetate is the most commonly used substrate for detecting the catalytic activity of esterases, including those that activate prodrugs in human cells. This substrate is unstable in aqueous solution, limiting its utility. Here, a stable chromogenic substrate for esterases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service