Skip to Content
Merck
All Photos(1)

Key Documents

754633

Sigma-Aldrich

(R)-(4,4′,6,6′-Tetramethoxybiphenyl-2,2′-diyl) bis{bis[3,5-bis(trifluoromethyl)phenyl]phosphine}

97%

Synonym(s):

(R)-(4,4′,6,6′-Tetramethoxy-1,1′-biphenyl-2,2′-diyl)bis(bis(3,5-bis(trifluoromethyl)phenyl)phosphine), (R)-BTFM-Garphos

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€329.00

€329.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€329.00

About This Item

Empirical Formula (Hill Notation):
C48H28F24O4P2
Molecular Weight:
1186.64
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

€329.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

optical activity

[α]22/D +33.0°, c = 0.5% in chloroform

mp

123-128 °C

functional group

fluoro
phosphine

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)P(c3c(c(cc(c3)OC)OC)c4c(cc(cc4OC)OC)P(c6cc(cc(c6)C(F)(F)F)C(F)(F)F)c5cc(cc(c5)C(F)(F)F)C(F)(F)F)c2cc(cc(c2)C(F)(F)F)C(F)(F)F

InChI

1S/C48H28F24O4P2/c1-73-29-17-35(75-3)39(37(19-29)77(31-9-21(41(49,50)51)5-22(10-31)42(52,53)54)32-11-23(43(55,56)57)6-24(12-32)44(58,59)60)40-36(76-4)18-30(74-2)20-38(40)78(33-13-25(45(61,62)63)7-26(14-33)46(64,65)66)34-15-27(47(67,68)69)8-28(16-34)48(70,71)72/h5-20H,1-4H3

InChI key

JDKXXRZDVSJSNQ-UHFFFAOYSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.
Garphos is a trademark of Kanata Chemical Technologies, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service