Skip to Content
Merck
All Photos(1)

Key Documents

753483

Sigma-Aldrich

5-Bromo-3-fluoropyridine-2-carboxylic acid

95%

Synonym(s):

5-Bromo-3-fluoropicolinic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€56.10

€56.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€56.10

About This Item

Empirical Formula (Hill Notation):
C6H3BrFNO2
CAS Number:
Molecular Weight:
220.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€56.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

175-180 °C

functional group

bromo
carboxylic acid
fluoro

SMILES string

OC(=O)c1ncc(Br)cc1F

InChI

1S/C6H3BrFNO2/c7-3-1-4(8)5(6(10)11)9-2-3/h1-2H,(H,10,11)

InChI key

JXHUXOAJAYWMHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service