Skip to Content
Merck
All Photos(1)

Key Documents

752975

Sigma-Aldrich

Ethyl 2-(hydroxymethyl)acrylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€148.00

€148.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€148.00

About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€148.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.450

density

1.068 g/mL at 25 °C

functional group

ester
hydroxyl

storage temp.

2-8°C

SMILES string

CCOC(=O)C(=C)CO

InChI

1S/C6H10O3/c1-3-9-6(8)5(2)4-7/h7H,2-4H2,1H3

InChI key

SYGAXBISYRORDR-UHFFFAOYSA-N

Related Categories

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

214.0 °F

Flash Point(C)

101.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cassandra L Petrou et al.
Journal of materials chemistry. B, 8(31), 6814-6826 (2020-04-29)
Fibrotic disorders account for over one third of mortalities worldwide. Despite great efforts to study the cellular and molecular processes underlying fibrosis, there are currently few effective therapies. Dual-stage polymerization reactions are an innovative tool for recreating heterogeneous increases in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service