Skip to Content
Merck
All Photos(1)

Key Documents

752223

Sigma-Aldrich

Diiodo(pentamethylcyclopentadienyl)iridium(III) dimer

Synonym(s):

di-μ-iododiiodobis[(1,2,3,4,5-η)-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl]di-iridium

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€97.40
1 G
€290.00

€97.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€97.40
1 G
€290.00

About This Item

Empirical Formula (Hill Notation):
C20H30I4Ir2
CAS Number:
Molecular Weight:
1162.50
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€97.40


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: iridium
reagent type: catalyst

mp

412-420 °C

storage temp.

2-8°C

SMILES string

I[Ir]I.I[Ir]I.C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.4HI.2Ir/c2*1-6-7(2)9(4)10(5)8(6)3;;;;;;/h2*1-5H3;4*1H;;/q;;;;;;2*+2/p-4

InChI key

GVAFOFZYINFTHJ-UHFFFAOYSA-J

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service