Skip to Content
Merck
All Photos(1)

Key Documents

751871

Sigma-Aldrich

3,4-Dihexylthiophene

97%

Synonym(s):

3,4-Bis-n-hexyl thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28S
CAS Number:
Molecular Weight:
252.46
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.494

density

0.925 g/mL at 25 °C

SMILES string

CCCCCCc1cscc1CCCCCC

InChI

1S/C16H28S/c1-3-5-7-9-11-15-13-17-14-16(15)12-10-8-6-4-2/h13-14H,3-12H2,1-2H3

InChI key

YIRIIAIZQBBXHL-UHFFFAOYSA-N

Application

This material is used in the synthesis of donor-acceptor copolymers[1] and low-band gap polymers[2] for use in Organic Photovoltaic Applications. It also has shown to enhance the thermal stability of polythiophene: fullerene solar cells.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Sivula et al.
Journal of the American Chemical Society, 128(43), 13988-13989 (2006-10-26)
Regioregularity has been shown to be an important aspect for the properties of poly(3-hexylthiophene) both in the pristine material and in bulk heterojunction photovoltaics with the fullerene derivative PCBM. Here we present a straightforward method to adjust the effective regioregularity
Banishoeib, F.; Henckens, A.; Fourier, S.; Vanhooyland, G.; Breselge, M.
Thin Solid Films, 516, 3978-3978 (2008)
Li, Y.; Xue, L.; Li, H.; Li, Z.; Xu, B.; Wen, S.; Tian, W.
Macromolecules, 42, 4491-4491 (2009)

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service