Skip to Content
Merck
All Photos(1)

Key Documents

750735

Sigma-Aldrich

3,5-Di-tert-butylphenol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€200.00
500 MG
€748.00

€200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€200.00
500 MG
€748.00

About This Item

Linear Formula:
[(CH3)3C]2C6H3OH
CAS Number:
Molecular Weight:
206.32
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

€200.00


Check Cart for Availability

Request a Bulk Order

form

powder

mp

87-89 °C (lit.)
90-91 °C

SMILES string

CC(C)(C)c1cc(O)cc(c1)C(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-7-11(14(4,5)6)9-12(15)8-10/h7-9,15H,1-6H3

InChI key

ZDWSNKPLZUXBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service