Skip to Content
Merck
All Photos(2)

Key Documents

750042

Sigma-Aldrich

1,3,5-Tris(2-thienyl)benzene

97%

Synonym(s):

1,3,5-Tri(thiophen-2-yl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12S3
CAS Number:
Molecular Weight:
324.48
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

154-160 °C

SMILES string

c1csc(c1)-c2cc(cc(c2)-c3cccs3)-c4cccs4

InChI

1S/C18H12S3/c1-4-16(19-7-1)13-10-14(17-5-2-8-20-17)12-15(11-13)18-6-3-9-21-18/h1-12H

InChI key

UBHPRZXDFVCNHZ-UHFFFAOYSA-N

General description

1,3,5-Tris(2-thienyl)benzene is a 1,3,5 tris substituted benzene that has a three dimensional structure which facilitates the preparation of conjugating polymers. It is a branched conductive monomer with electronically attached nodes that provide a suitable support to increase the conductivity of the synthesized polymers.

Application

1,3,5-Tris(2-thienyl)benzene is a trifunctionalized monomer that can be used in the synthesis of conjugated C3-symmetric poly(arylbenzene) polymers. These polymeric materials can further be used in organic photovoltaic devices as well as organic field effect transistors (OFETs) and organic light emitting diodes (OLED) systems.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical and spectral properties of meta-linked 1, 3, 5-tris (aryl) benzenes and 2, 4, 6-tris (aryl)-1-phenoles, and their polymers
Idzik KR, et al.
Electrochimica Acta, 55(24), 7419-7426 (2010)
Synthesis and Electrochemical Properties of Novel 1, 3, 5-Tris (oligothienyl) benzenes: A New Generation of 3D Reticulating Agents
Cherioux F and Guyard L
Advances in Functional Materials, 11(4), 305-309 (2001)
Shape and size effects in the crystal structures of complexes of 1, 3, 5-trinitrobenzene with some trigonal donors: the benzene-thiophene exchange rule
Thallapally PK, et al.
Tetrahedron, 56(36), 6721-6728 (2000)
Idzik, K. R.; Beckert, R.; Golba, S.; Ledwon, P.; Lapkowski, M.
Tetrahedron Letters, 51, 2396-2396 (2010)
Synthesis of C3-Symmetric Nano-Sized Polyaromatic Compounds by Trimerization and Suzuki- Miyaura Cross-Coupling Reactions
Kotha S, et al.
European Journal of Organic Chemistry, 2004(19), 4003-4013 (2004)

Articles

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service