Skip to Content
Merck
All Photos(1)

Key Documents

748722

Sigma-Aldrich

N,N′-Dimethyl-1,2-phenylenediamine

97%

Synonym(s):

N,N′-Dimethyl-1,2-diaminobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€125.00

€125.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€125.00

About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€125.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

97%

form

solid

refractive index

n20/D 1.592

density

1.262 g/mL at 25 °C

functional group

amine

SMILES string

CNc1ccccc1NC

InChI

1S/C8H12N2/c1-9-7-5-3-4-6-8(7)10-2/h3-6,9-10H,1-2H3

InChI key

UMAPFAAAQBMYNJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin L Willis et al.
PloS one, 6(7), e21159-e21159 (2011-07-19)
After ovulation, non-pregnant female giant pandas experience pseudopregnancy. During pseudopregnancy, non-pregnant females exhibit physiological and behavioral changes similar to pregnancy. Monitoring hormonal patterns that are usually different in pregnant mammals are not effective at determining pregnancy status in many animals

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service