Skip to Content
Merck
All Photos(3)

Documents

748005

Sigma-Aldrich

Gold nanorods

10 nm diameter, silica coated, λmax, 850 nm, dispersion in H2O

Synonym(s):

Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

material

silica

description

Silica coated Gold nanorod
Silica coating thickness 18-22 nm (TEM)

form

dispersion in H2O
nanorod

concentration

>30 μg/mL in H2O

diam. × L

9-11 nm × 41-48 nm

color

purple

diameter

10 nm

density

0.995 g/mL at 25 °C

λmax

850 nm

storage temp.

2-8°C

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Our silica coated gold nanorods are highly stable and suitable for applications including; photothermal therapy; fluorescence enhancement; plasmonic arrays; and photoacoustic amplifiers.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Sheng Chen et al.
Nano letters, 11(2), 348-354 (2011-01-20)
Photoacoustic signal generation by metal nanoparticles relies on the efficient conversion of light to heat, its transfer to the environment, and the production of pressure transients. In this study we demonstrate that a dielectric shell has a strong influence on
Stéphanie Vial et al.
Langmuir : the ACS journal of surfaces and colloids, 23(8), 4606-4611 (2007-03-21)
A systematic study of the optical effects derived from plasmon coupling in mono- and multilayers of gold nanorods is presented. The monolayers were prepared using the standard polyelectrolyte-assisted layer-by-layer (LbL) method and gold nanorods coated with either poly(N-vinyl pyrrolidone) or
Liu; Si-Yun;
The Journal of Physical Chemistry C, 117(20), 10636-10642 (2013)
Sudipta Mallick et al.
Journal of nanoscience and nanotechnology, 13(5), 3223-3229 (2013-07-19)
Due to their efficient conversion of absorbed light energy to heat gold nanorods have been proved to be an amazing tool for minimally invasive photo-thermal cancer therapy. The present in vitro study demonstrates the ability of silica coated Au nanorods

Articles

Gold nanoparticles find diverse applications from biomedical engineering to photovoltaics, leveraging their tunable optical properties for various fields.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service