Skip to Content
Merck
All Photos(1)

Key Documents

746932

Sigma-Aldrich

Polystyrene thiol terminated

average Mn 11,000, PDI ≤1.1

Synonym(s):

MercaptoPS, Mercaptopolystyrene, PS SH terminated

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€246.00

€246.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€246.00

About This Item

Linear Formula:
CNC3H6(C8H8)nSH
UNSPSC Code:
12352100
NACRES:
NA.23

€246.00


Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average Mn 11,000

mp

254-341 °C (decomposition)

transition temp

Tg 99 °C (onset)

PDI

≤1.1

Looking for similar products? Visit Product Comparison Guide

Application

This is a well-defined polystyrene with thiol functionality on one end and cyano on the other. The thiol functionality is veratile and commonly used in surface modifying nanoparticles; attaching to Au to form self assembled monolayers (SAMs); as well as thiol - ene click chemistry.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents 7; 714; 075; 7; 250; 479; 7; 666; 962; 6; 642; 318; 6; 747; 111; W2010/8356.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service