Skip to Content
Merck
All Photos(1)

Key Documents

746029

Sigma-Aldrich

5-Fluoroindoline

97%

Synonym(s):

5-Fluoro-2,3-dihydro-1H-indole, 5-Fluoro-2,3-dihydroindole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€92.40

€92.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€92.40

About This Item

Empirical Formula (Hill Notation):
C8H8FN
CAS Number:
Molecular Weight:
137.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€92.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n/D 1.556

density

1.171 g/mL at 25 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

Fc1cc2c(cc1)NCC2

InChI

1S/C8H8FN/c9-7-1-2-8-6(5-7)3-4-10-8/h1-2,5,10H,3-4H2

InChI key

NXQRMQIYCWFDGP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

225.1 °F

Flash Point(C)

107.25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service