Skip to Content
Merck
All Photos(1)

Key Documents

745413

Sigma-Aldrich

Potassium benzyl cyanocarbamate

Synonym(s):

Looper Reagent, Potassium N-cyanocarbamic acid, phenylmethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€268.00

€268.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€268.00

About This Item

Empirical Formula (Hill Notation):
C9H7KN2O2
CAS Number:
Molecular Weight:
214.26
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€268.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

228-232 °C

functional group

amine
phenyl

SMILES string

[K]N(C#N)C(=O)OCc1ccccc1

InChI

1S/C9H8N2O2.K/c10-7-11-9(12)13-6-8-4-2-1-3-5-8;/h1-5H,6H2,(H,11,12);/q;+1/p-1

InChI key

HZBHQPNPJVDRMR-UHFFFAOYSA-M

Application

In tandem with TMSCl, product generates an in situ N-silylcarbodiimide that rapidly guanylates a variety of amines at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Looper; R. E.;
The Journal of Organic Chemistry, 76, 6957-6957 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service