Skip to Content
Merck
All Photos(1)

Key Documents

744484

Sigma-Aldrich

Vinyl chloride

≥99.95%

Synonym(s):

Chloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCl
CAS Number:
Molecular Weight:
62.50
Beilstein:
1731576
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

Assay

≥99.95%

form

gas

refractive index

n20/D 1.3700 (lit.)

bp

-13.4 °C (lit.)

mp

-153.8 °C (lit.)

density

0.911 g/mL at 25 °C (lit.)

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vinyl chloride can be used as a monomer precursor:
  • In the living radical polymerization reaction using copper catalysts.
  • In the synthesis of polymer poly(vinyl chloride) (PVC) , poly(vinyl chloride)/montmorillonite (PVC/MMT) nanocomposites by intercalative polymerization process.

Packaging

500ml high pressure aluminum cylinder equipped with a stainless steel valve according to DIN 477-1.

Other Notes

For 744484-400G-EU packaging: 1000 mL of steel cylinder equipped with stainless steel valve.

Recommended products

For 744484-400G-EU: outlet fitting: DIN 477-1 no. 1, Z742162 or Z742164 are the recommended regulators.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-79.6 °F - closed cup

Flash Point(C)

-62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous room temperature metal-catalyzed living radical polymerization of vinyl chloride
Percec V, et al.
Journal of the American Chemical Society, 124(18), 4940-4941 (2002)
Synthesis and structure of poly (vinyl chloride)
Endo K
Progress in Polymer Science, 27(10), 2021-2054 (2002)
Thermal properties of poly (vinyl chloride)/montmorillonite nanocomposites
Gong F, et al.
Polymer Degradation and Stability, 84(2), 289-294 (2004)
Vinyl chloride monomer and hepatocarcinoma.
N Bourgeois
Acta gastro-enterologica Belgica, 64(1), 25-34 (2001-04-27)
J Kielhorn et al.
Environmental health perspectives, 108(7), 579-588 (2000-07-25)
Vinyl chloride (VC) is both a known carcinogen and a regulated chemical, and its production capacity has almost doubled over the last 20 years, currently 27 million tons/year worldwide. According to recent reports it is still a cause for concern.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service