Skip to Content
Merck
All Photos(1)

Key Documents

744441

Sigma-Aldrich

5-Chloro-5H-dibenzo[a,d]cycloheptene

97%

Synonym(s):

5H-Dibenzo[a,d]cyclohepten-5-yl chloride, Dibenzosuberenyl chloride, tropCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl
CAS Number:
Molecular Weight:
226.70
Beilstein:
515093
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥96.5% (HPLC)
97%

form

powder

SMILES string

ClC1C(C=CC=C2)=C2C=CC3=C1C=CC=C3

InChI

1S/C15H11Cl/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-10,15H

InChI key

XUDMFUCJNFYMRZ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electromeric rhodium radical complexes.
Florian Frank Puschmann et al.
Angewandte Chemie (International ed. in English), 49(2), 385-389 (2009-12-04)
Chiral Rhodium(I) and Iridium(I) Amino-Olefin Complexes:? pKa, N-H Bond Dissociation Energy, and Catalytic Transfer Hydrogenation
Maire, P., et al.
Organometallics, 24, 3207-3218 (2005)
Torsten Büttner et al.
Chemical communications (Cambridge, England), 24(24), 2820-2821 (2004-12-16)
A combination of the rigid bis(5H-dibenzo[a,d]cyclohepten-5-yl)amine (trop(2)NH) and 5-amino-5H-dibenzo[a,d]cycloheptene (tropNH(2)) ligand allowed the synthesis of the stable pentacoordinated 18 electron amine olefin rhodium(i) complex [Rh(trop(2)NH)(tropNH(2))]O(3)SCF(3)(); this complex can be cleanly deprotonated [pK(a)(DMSO) = 20.6(1)] to the corresponding amide [Rh(trop(2)N)(tropNH(2))](6) which
Ethanol as hydrogen donor: highly efficient transfer hydrogenations with rhodium(I) amides.
Theo Zweifel et al.
Angewandte Chemie (International ed. in English), 47(17), 3245-3249 (2008-03-19)
Heterolytic splitting of hydrogen with rhodium(I) amides.
Pascal Maire et al.
Angewandte Chemie (International ed. in English), 44(39), 6318-6323 (2005-09-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service