Skip to Content
Merck
All Photos(1)

Key Documents

743704

Sigma-Aldrich

1,2-Butadiene

≥97.0%

Synonym(s):

1-Methylallene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C=CHCH3
CAS Number:
Molecular Weight:
54.09
Beilstein:
1730808
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

gas

bp

10.8 °C (lit.)

SMILES string

CC=C=C

InChI

1S/C4H6/c1-3-4-2/h4H,1H2,2H3

InChI key

QNRMTGGDHLBXQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Butadiene can be used as a reagent:
  • To synthesize useful β-borylallylsilanes building blocks by reacting with various achiral silylboranes via Pd-catalyzed asymmetric silaboration reaction.[1]
  • In the enantioselective diboration reaction in the presence of palladium catalyst and Lewis base ligand.[2]
  • 3-methyl-4-methyleneisocoumarin via formation of ortho-thallated benzoic acid in the presence of thallium trifluoroacetate and palladium catalyst.[3]

Packaging

500ml high pressure aluminum cylinder equipped with a stainless steel valve according to DIN 477-1.

Other Notes

For 743704-250g-EU packaging: 500 mL of aluminium cylinder equipped with brass valve.

Recommended products

For 743704-250g-EU: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742161 are the recommended regulators.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of isocoumarins via thallation-olefination of benzoic acids
Larock RC, et al.
Journal of the American Chemical Society, 106(18), 5274-5284 (1984)
Palladium-catalyzed enantioselective diboration of prochiral allenes
Pelz NF, et al.
Journal of the American Chemical Society, 126(50), 16328-16329 (2004)
Palladium-catalyzed asymmetric silaboration of allenes.
Ohmura T, et al.
Journal of the American Chemical Society, 128(42), 13682-13683 (2006)
Ralf I Kaiser et al.
The journal of physical chemistry. A, 118(32), 6181-6190 (2014-08-02)
The reactions of the 4-tolyl radical (C6H4CH3) and of the D7-4-tolyl radical (C6D4CD3) with 1,2-butadiene (C4H6) have been probed in crossed molecular beams under single collision conditions at a collision energy of about 54 kJ mol(-1) and studied theoretically using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service