Skip to Content
Merck
All Photos(1)

Key Documents

742945

Sigma-Aldrich

TurboBeads TEMPO

≥99%

Synonym(s):

Nano particles, magnetic, TEMPO functionalized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352128
NACRES:
NA.22

product line

TurboBeads

Assay

≥99%

form

powder

composition

carbon content, ≤14 wt. %

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

extent of labeling

≥0.1 mmol/g loading (TEMPO)

magnetization

≥120 emu/g, mass saturation

surface area

≥15 m2/g

average diameter

≤50 nm

suitability

conforms to structure for Infrared spectrum

Application

Recyclable TEMPO (2,2,6,6-tetramethylpiperidine 1-oxyl radical)-coated nanoparticles have been prepared by the immobilization of TEMPO on iron oxide (Fe3O4) superparamagnetic nanoparticles.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

Proof of identity: metallic cobalt identity from XRD

air-stability:
weight gain in air at 400°C >20 wt.%
weight gain in air at 100°C <3 wt.%

Legal Information

TurboBeads is a trademark of TurboBeads LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander K Tucker-Schwartz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(42), 12718-12726 (2010-09-21)
The organic oxidant TEMPO (2,2,4,4-tetramethylpiperdine-1-oxyl) was immobilized on iron oxide (Fe(3)O(4)) superparamagnetic nanoparticles by employing strong metal-oxide chelating phosphonates and azide/alkyne "click" chemistry. This simple preparation yields recyclable TEMPO-coated nanoparticles with good TEMPO loadings. They have excellent magnetic response and
Saroj Kumar et al.
Journal of nanobiotechnology, 11, 14-14 (2013-05-04)
Introduction of effective point-of-care devices for use in medical diagnostics is part of strategies to combat accelerating health-care costs. Molecular motor driven nanodevices have unique potentials in this regard due to unprecedented level of miniaturization and independence of external pumps.
Vladimir Zlateski et al.
Bioconjugate chemistry, 25(4), 677-684 (2014-03-29)
In the pursuit of robust and reusable biocatalysts for industrial synthetic chemistry, nanobiotechnology is currently taking a significant part. Recently, enzymes have been immobilized on different nanoscaffold supports. Carbon coated metallic nanoparticles were found to be a practically useful support
Inge K Herrmann et al.
Nanoscale, 5(18), 8718-8723 (2013-08-01)
In a number of clinical conditions such as intoxication, bacteraemia or autoimmune diseases the removal of the disease-causing factor from blood would be the most direct cure. However, physicochemical characteristics of the target compounds limit the applicability of classical filtration
Quirin M Kainz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(30), 10038-10045 (2013-06-19)
Unprecedented magnetic borohydride exchange (mBER), magnetic Wang aldehyde (mWang) and magnetic amine resins were prepared from highly magnetic polymer-coated cobalt or iron nanoparticles. Microwave irradiation was used to obtain excellent degrees of functionalization (>95 %) and loadings (up to 3.0 mmol  g(-1))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service