Skip to Content
Merck
All Photos(1)

Key Documents

741000

Sigma-Aldrich

Tin(II) sulfide

granular, ≥99.99% trace metals basis

Synonym(s):

Herzenbergite, Stannous sulfide, Tin monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SSn
CAS Number:
Molecular Weight:
150.78
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

granular

density

5.22 g/mL at 25 °C (lit.)

SMILES string

S=[SnH2]

InChI

1S/S.Sn

InChI key

AFNRRBXCCXDRPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organotin unsymmetric dithiocarbamates: synthesis, formation and characterisation of tin (II) sulfide films by atmospheric pressure chemical vapour deposition.
Kana AT, et al.
Polyhedron, 20(24), 2989- 2995 (2001)
Miles, R. W.; Ogah, O. E.; Zoppi, G.; Forbes, I.
Thin Solid Films, 517, 4702-4702 (2009)
Jiang, T. and Ozin, G. A.
Journal of Materials Chemistry, 8, 1099-1099 (1998)
Parker, D. and Singh D. J.
Journal of Applied Physics, 108, 083712-083712 (2010)
Antimony-doped tin (II) sulfide thin films.
Sinsermsuksakul P, et al.
Chemistry of Materials, 24(23), 4556-4562 (2012)

Articles

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service