Skip to Content
Merck
All Photos(1)

Key Documents

739383

Sigma-Aldrich

2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde

97%

Synonym(s):

2,2-Difluoro-1,3-benzodioxole-4carbaldehyde, 2,2-Difluorobenzo[d][1,3]dioxole, 4-Formyl-2,2-difluoro-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€68.70

€68.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€68.70

About This Item

Empirical Formula (Hill Notation):
C7H4F2O2
CAS Number:
Molecular Weight:
158.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€68.70


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.497

density

1.423 g/mL at 25 °C

functional group

aldehyde
fluoro

storage temp.

−20°C

SMILES string

FC1(F)Oc2cccc(C=O)c2O1

InChI

1S/C8H4F2O3/c9-8(10)12-6-3-1-2-5(4-11)7(6)13-8/h1-4H

InChI key

NIBFJPXGNVPNHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

196.0 °F

Flash Point(C)

91.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service